o-Anisidine ≥99%

Danger

Synonyms: 2-Anisidine, 2-Aminoanisole, 2-Methoxyaniline

A88182-500G A88182-100G A88182-5G
220105-182EA 122.77 USD
220105-182 220105-181 220105-183
o-Anisidine ≥99%
o-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007688
CAS Number: 90-04-0
EINECS: 201-963-1
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR