α-Angelicalactone

Synonyms: 5-Methylfuran-2(3H)-one

A86406-100G A86406-10G
220105-105EA 228.31 USD
220105-105 220105-106
α-Angelicalactone
α-Angelicalactone
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.0904 g/cm³ (12 °C)
MDL Number: MFCD00005375
CAS Number: 591-12-8
EINECS: 209-701-8
Merck Index: 12,00686

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR