Adenine

Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A8626-1KG A8626-100G A8626-1G A8626-25G A8626-5G
220105-102EA 2976.92 USD
220105-102 220105-100 220105-101 220105-103 220105-104
Adenine
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR