Aniline hydrochloride

Danger

Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride

A8524-5G A8524-50G
220105-009EA 64.18 USD
220105-009 220105-008
Aniline hydrochloride
Aniline hydrochloride
Formula: C₆H₈ClN
MW: 129.59 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 198 °C
Density: 1.2215 g/cm³ (20 °C)
Flash Pt: 193 °C
MDL Number: MFCD00012958
CAS Number: 142-04-1
EINECS: 205-519-8
UN: 1548
ADR: 6.1,III
Merck Index: 13,00616

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR