Lithium acetoacetate

Synonyms: 3-Oxobutyric acid Lithium salt, Lithium 3-Oxobutyrate, Acetoacetic acid Lithium salt

A8509-1G A8509-10MG A8509-5G A8509-250MG
220102-875EA 126 USD
220102-875 220102-874 220103-354 220102-876
Lithium acetoacetate
Lithium acetoacetate
Formula: C₄H₅LiO₃
MW: 108.02 g/mol
MDL Number: MFCD00013060
CAS Number: 3483-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR