L(+)-Asparagine monohydrate

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

A8381-1KG A8381-100G
220104-659EA 564.92 USD
220104-659 220104-658
L(+)-Asparagine monohydrate
L(+)-Asparagine monohydrate
Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,00837

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR