Acetohydrazide ≥90%

Warning

Synonyms: Acetic hydrazide, Acethydrazide

A8309-100G A8309-25G A8309-5G
220104-921EA 370.46 USD
220104-921 220104-922 220104-923
Acetohydrazide ≥90%
Acetohydrazide
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 63…65 °C
Flash Pt: >100 °C
MDL Number: MFCD00007610
CAS Number: 1068-57-1
EINECS: 213-948-7

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR