3,4-Dimethoxyaniline ≥98%

Warning

A83008-25G A83008-10G
220104-919EA 191.69 USD
220104-919 220104-918
3,4-Dimethoxyaniline ≥98%
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 85…88 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR