11-Aminoundecanoic acid ≥97%

Synonyms: 11-Aminoundecylic acid

A82605-100G A82605-500G
220104-906EA 69.78 USD
220104-906 220104-907
11-Aminoundecanoic acid ≥97%
11-Aminoundecanoic acid
Formula: C₁₁H₂₃NO₂
MW: 201.31 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 187…190 °C
MDL Number: MFCD00008150
CAS Number: 2432-99-7
EINECS: 219-417-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR