4-Amino-4H-1,2,4-triazole

Synonyms: 4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine

A81803-25G A81803-100G A81803-5G
220102-724EA 46.85 USD
220102-724 220102-723 220102-725
4-Amino-4H-1,2,4-triazole
4-Amino-4H-1,2,4-triazole
Formula: C₂H₄N₄
MW: 84.08 g/mol
MDL Number: MFCD00003099
CAS Number: 584-13-4
EINECS: 209-533-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR