5-Aminosalicylic acid ≥95%

A79809-5G A79809-100G
220104-777EA 44.8 USD
220104-777 220104-776
5-Aminosalicylic acid ≥95%
5-Aminosalicylic acid
Formula: C₇H₇NO₃
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 280 °C
Density: 1.568 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR