5-Aminoquinoline ≥97%

Warning

A79205-1G A79205-5G
220104-750EA 84.97 USD
220104-750 220104-751
5-Aminoquinoline ≥97%
5-Aminoquinoline
Formula: C₉H₈N₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7
EINECS: 210-266-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR