4-Amino-2-methylquinoline ≥98%

Warning

Synonyms: 4-Aminoquinaldine, 2-Methylquinolin-4-amine

A79000-25G A79000-5G
220104-737EA 340.31 USD
220104-737 220104-738
4-Amino-2-methylquinoline ≥98%
4-Amino-2-methylquinoline
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00006759
CAS Number: 6628-04-2
EINECS: 229-604-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR