2-Aminopyrimidine

Synonyms: Pyrimidin-2-ylamine, Pyrimidin-2-amine

A78608-100G A78608-500G
220104-726EA 89.28 USD
220104-726 220104-727
2-Aminopyrimidine
2-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR