3-Aminopyridine

Danger

Synonyms: Pyridin-3-amine, 3-Pyridylamine

A78209-100G A78209-500G A78209-25G
220104-709EA 148.62 USD
220104-709 220104-711 220104-710
3-Aminopyridine
3-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: ∼248 °C (1013 hPa)
Melting Pt: 60…63 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR