2-Aminopyridine

Synonyms: 2-Pyridylamine, Pyridin-2-amine

A77989-25G A77989-100G
220104-643EA 58.69 USD
220104-643 220104-642
2-Aminopyridine
2-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 210.6 °C (1013 hPa)
Melting Pt: 50 - 52 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR