Sodium L(+)-ascorbate

Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

A7631-1KG A7631-25G A7631-500G A7631-100G
220104-568EA 316.4 USD
220104-568 220104-569 220104-570 220104-567
Sodium L(+)-ascorbate
Sodium L(+)-ascorbate
Formula: C₆H₇NaO₆
MW: 198.11 g/mol
Melting Pt: 219 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
EINECS: 205-126-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR