L-Alanine ≥98%

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

A7627-500G A7627-1G A7627-1KG A7627-100G
220104-566EA 499.69 USD
220104-566 220104-564 220104-565 220104-563
L-Alanine ≥98%
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR