2-Amino-6-methylpyridine

Danger

Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline

A75706-100G A75706-500G
220104-540EA 65.05 USD
220104-540 220104-541
2-Amino-6-methylpyridine
2-Amino-6-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 214.4 °C (1013 hPa)
Melting Pt: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00006331
CAS Number: 1824-81-3
EINECS: 217-360-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR