2-Amino-5-methylpyridine

Synonyms: 6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline

A75684-50G A75684-10G
220104-539EA 100.58 USD
220104-539 220104-538
2-Amino-5-methylpyridine
2-Amino-5-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 227.1 °C (1013 hPa)
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00006328
CAS Number: 1603-41-4
EINECS: 216-503-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR