2-Amino-3-methylpyridine

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

A75633-100G A75633-500G
220104-536EA 98.65 USD
220104-536 220104-537
2-Amino-3-methylpyridine
2-Amino-3-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221.1 °C (1013 hPa)
Melting Pt: 29 - 32 °C
Density: 1.073 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR