Dapsone ≥97%

Warning

Synonyms: 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone

A74807-100G A74807-500G
220104-501EA 99.77 USD
220104-501 220104-502
Dapsone ≥97%
Dapsone
Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
MDL Number: MFCD00007887
CAS Number: 80-08-0
EINECS: 201-248-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR