D-Alanine ≥98%

Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine

A7377-100G A7377-25G A7377-5G
220104-488EA 749.54 USD
220104-488 220104-489 220104-490
D-Alanine ≥98%
D-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR