(4-Aminophenyl)acetic acid ≥98%

A71352-100G A71352-25G
220104-410EA 326.31 USD
220104-410 220104-411
(4-Aminophenyl)acetic acid ≥98%
(4-Aminophenyl)acetic acid
Formula: (C₆H₄(NH₂))CH₂COOH
MW: 151.16 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 199…202 °C
MDL Number: MFCD00007916
CAS Number: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR