2-Amino-4-nitrophenol ≥96%

Warning

Synonyms: 2-Hydroxy-5-nitroaniline

A70402-5G A70402-25G
220104-390EA 50.08 USD
220104-390 220104-389
2-Amino-4-nitrophenol ≥96%
2-Amino-4-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 176 - 177 °C
Density: 1.66 g/cm³ (20 °C)
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR