Alginic acid (from Macrocystis pyrifera (kelp))

A7003-100G A7003-250G A7003-500G A7003-1KG
220104-351EA 86.38 USD
220104-351 220104-353 220104-354 220104-352
Alginic acid (from Macrocystis pyrifera (kelp))
Alginic acid

Alginic acid is a high molecular weight linear polymer isolated from seaweed. It is a linear β-1→4-linked polymer of n-mannuronic and L-guluronic acid. It is an acid polysaccharide present in the extracellular matrix of brown algae.


It is reported to inhibit the reverse transcriptase activity of HIV. Structural investigation of alginic acid, a marine algal polysaccharide poly-α-L-guluronic acid, has been reported. Anti-inflammatory effect of alginic acid, isolated from the brown algae Sargassum wightii, in type II collagen induced arthritic rats has been reported. Pyrolysis of alginic acid using py-GC/MS over a wide temperature range (200-800°C) and thermogravimetric analysis (TGA) has been reported. It was reported to cause the biocorrosion of thin films of copper on germanium internal reflection elements (IREs).


Alginic acid is suitable reagent used to study the in-vitro and in-vivo mitogenic activity of alginates. It is suitable for use in the spectrophotometric determination of transparent exopolymer particles by dye-binding assay

Formula: (C₆H₈O₆)n
MW: 176.13 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081309
CAS Number: 9005-32-7
EINECS: 232-680-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea02d0735fded7cad93ac53c779092579becba4&vid=765890130956b20ed9fae7c669554c5198c517f5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR