2-Aminonicotinic acid

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

A68300-25G A68300-5G
220104-272EA 118.46 USD
220104-272 220104-273
2-Aminonicotinic acid
2-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 297 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS Number: 5345-47-1
EINECS: 226-296-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR