4-(Aminomethyl)pyridine

Synonyms: 4-Picolamine, (4-Pyridyl)methylamine, Pyridin-4-ylmethanamine, 4-Picolylamine, 4-Pyridylmethylamine

A65603-5G A65603-25G
220104-194EA 43.29 USD
220104-194 220104-193
4-(Aminomethyl)pyridine
4-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.0688 g/cm³ (20 °C)
MDL Number: MFCD00006449
CAS Number: 3731-53-1
EINECS: 223-092-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR