3-(Aminomethyl)pyridine

Synonyms: 3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine

A65409-100G A65409-5G
220103-942EA 78.62 USD
220103-942 220103-943
3-(Aminomethyl)pyridine
3-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.0712 g/cm³ (20 °C)
MDL Number: MFCD00006412
CAS Number: 3731-52-0
EINECS: 223-091-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR