2-(Aminomethyl)pyridine

Synonyms: 2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine

A65204-5G A65204-100G A65204-25G
220103-933EA 28.11 USD
220103-933 220103-931 220103-932
2-(Aminomethyl)pyridine
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 81 °C
Density: 1.0583 g/cm³ (20 °C)
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR