N-Acetyl-DL-tryptophan

Synonyms: Ac-DL-Trp-OH, N-Acetyl-DL-tryptophan

A6251-1KG A6251-5G A6251-25G
220104-025EA 1723.08 USD
220104-025 220104-027 220104-026
N-Acetyl-DL-tryptophan
N-Acetyl-DL-tryptophan
Formula: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
Melting Pt: 205…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00005644
CAS Number: 87-32-1
EINECS: 201-739-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR