3-Methylanthranil acid ≥99%

Warning

Synonyms: 2-Amino-3-methylbenzoic acid

A62205-25G A62205-5G
220102-520EA 552.37 USD
220102-520 220102-521
3-Methylanthranil acid ≥99%
3-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 174 - 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00007745
CAS Number: 4389-45-1
EINECS: 224-505-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR