Ammonium hydrogen carbonate ≥99.0%, ReagentPlus®

Warning

Synonyms: Ammonium bicarbonate

A6141-25G A6141-1KG A6141-500G
220104-002EA 24.74 USD
220104-002 220104-001 220104-003
Ammonium hydrogen carbonate ≥99.0%, ReagentPlus®
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 35 °C
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR