5-Aminoindole

Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine

A59654-1G A59654-5G
220103-890EA 26.92 USD
220103-890 220103-891
5-Aminoindole
5-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 190 °C (6 mmHg)
Melting Pt: 131…134 °C
MDL Number: MFCD00005679
CAS Number: 5192-03-0
EINECS: 225-977-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR