6-Aminohexan-1-ol ≥97%

Warning

Synonyms: 6-Amino-1-hexanol

A56353-25G A56353-5G A56353-100G
220103-780EA 255.23 USD
220103-780 220103-781 220103-779
6-Aminohexan-1-ol ≥97%
6-Aminohexan-1-ol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 53…55 °C
MDL Number: MFCD00008241
CAS Number: 4048-33-3
EINECS: 223-748-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR