2-Amino-4,6-dihydroxypyrimidine

Synonyms: 2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one

A50401-100G A50401-25G
220102-235EA 956.31 USD
220102-235 220102-236
2-Amino-4,6-dihydroxypyrimidine
2-Amino-4,6-dihydroxypyrimidine
Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
Boiling Pt: 394 °C (1013 hPa)
Melting Pt: 300 °C
MDL Number: MFCD00006094
CAS Number: 56-09-7
EINECS: 200-256-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR