2-Amino-4,6-dichloropyrimidine

Synonyms: 4,6-Dichloropyrimidin-2-ylamine

A48601-100G A48601-10G
220102-783EA 287.54 USD
220102-783 220102-784
2-Amino-4,6-dichloropyrimidine
2-Amino-4,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
Melting Pt: 216…220 °C
MDL Number: MFCD00006090
CAS Number: 56-05-3
EINECS: 200-253-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR