3-Amino-2-chloropyridine

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

A46900-10G A46900-50G
220103-469EA 60.42 USD
220103-469 220103-470
3-Amino-2-chloropyridine
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 78 - 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR