2-Amino-5-chlorobenzophenone ≥98%

Synonyms: 1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone

A45564-100G A45564-25G
220103-145EA 66.34 USD
220103-145 220103-146
2-Amino-5-chlorobenzophenone ≥98%
2-Amino-5-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Boiling Pt: 424 °C (1013 hPa)
Melting Pt: 96…99 °C
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR