L(+)-Arginine ≥98.5%, crystalline powder Ph. Eur., USP, Ajinomoto cell culture reagent, GMP, SAFC®

Warning

A4474-1KG A4474-100G A4474-10KG A4474-25KG
220103-335EA 790.77 USD
220103-335 102365-576 102365-578 102365-580
L(+)-Arginine ≥98.5%, crystalline powder Ph. Eur., USP, Ajinomoto cell culture reagent, GMP, SAFC®
L(+)-Arginine

Manufactured and Packaged under cGMP.

Formula: C₆H₁₄N₄O₂
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR