4-Bromo-1-naphthylamine

Synonyms: 1-Amino-4-bromonaphthalene

A43405-5G A43405-25G
220103-286EA 141.08 USD
220103-286 220103-285
4-Bromo-1-naphthylamine
4-Bromo-1-naphthylamine
Formula: C₁₀H₈BrN
MW: 222.08 g/mol
MDL Number: MFCD00004023
CAS Number: 2298-07-9
EINECS: 218-944-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR