4-Aminobenzophenone

Synonyms: (4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone

A41402-25G A41402-10G
220103-214EA 138.92 USD
220103-214 220103-213
4-Aminobenzophenone
4-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR