2-Aminobenzophenone ≥98%

Synonyms: o-Aminobenzophenone

A41208-25G A41208-5G
220103-210EA 95.95 USD
220103-210 220103-211
2-Aminobenzophenone ≥98%
2-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 105…106 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR