1-Aminocyclopropanecarboxylic acid

A3903-250MG A3903-100MG A3903-1G
220102-846EA 270.37 USD
220102-846 220103-089 220103-090
1-Aminocyclopropanecarboxylic acid
1-Aminocyclopropanecarboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR