Alizarin complexone

Synonyms: Alizarin-3-methyliminodiacetic acid, 2,2'-(((3,4-Dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl)azanediyl)diacetic acid, 3,4-dihydroxyanthraquinon-2-ylmethyliminodi(acetic acid), Alizarine-3-methylamine-N,N-diacetic acid

A3882-1G A3882-10G A3882-5G
220103-080EA 247.69 USD
220103-080 220103-079 220103-081
Alizarin complexone
Alizarin complexone
Formula: C₁₉H₁₅NO₈
MW: 385.33 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00001202
CAS Number: 3952-78-1
EINECS: 223-544-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR