Ammonium nitrate

Warning

A3795-2.5KG A3795-25KG A3795-500G
220103-046EA 301.6 USD
220103-046 220103-047 220103-048
Ammonium nitrate
Ammonium nitrate
Formula: NH₄NO₃
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 169 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 13,00538

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR