Ammonium persulfate (APS) for electrophoresis

Danger

A3678-100G A3678-25G
220103-006EA 51.17 USD
220103-006 220103-007
Ammonium persulfate (APS) for electrophoresis
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR