Allyl bromide ≥99%, ReagentPlus®

Synonyms: 3-Bromopropene, 3-Propenyl bromide

A29585-1KG A29585-500G A29585-5G A29585-3KG A29585-100G
220102-553EA 163.69 USD
220102-553 220101-322 220101-323 220101-321 220102-552
Allyl bromide ≥99%, ReagentPlus®
Allyl bromide
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: –119 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: –1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000244
CAS Number: 106-95-6
EINECS: 203-446-6
UN: 1099
ADR: 3,I
Merck Index: 13,00285

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR