4-Phenylaniline

Danger

Synonyms: 4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl

A2898-25G A2898-1G A2898-5G
220102-465EA 37.62 USD
220102-465 220102-464 220102-466
4-Phenylaniline
4-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 55 °C
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00007879
CAS Number: 92-67-1
EINECS: 202-177-1
Merck Index: 12,01276

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR