Methyl hydrogen adipate ≥99%

Synonyms: 1,6-Hexanedioic acid methyl ester, Adipic acid methyl ester, Adipic acid monomethyl ester, 1,6-Hexanedioic acid monomethyl ester, Monomethyl adipate

A26403-5G A26403-25G
220101-835EA 60.95 USD
220101-835 220101-834
Methyl hydrogen adipate ≥99%
Methyl hydrogen adipate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 292 °C (1013 hPa)
Density: 1.1236 g/cm³ (20 °C)
MDL Number: MFCD00004418
CAS Number: 627-91-8
EINECS: 211-019-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR